Invitrogen
Manufacturer Code:PA5114204
Catalog # PIPA5114204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Mouse Laptm4b |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_14051 C330023P13Rik LAPTM4B LAPTM4beta LC27 LOC100356277 lysosomal associated protein transmembrane 4 beta lysosomal protein transmembrane 4 beta lysosomal-associated protein transmembrane 4B lysosomal-associated transmembrane protein 4 Lysosomal-associated transmembrane protein 4B Lysosomal-associated transmembrane protein 4B-like Lysosome-associated transmembrane protein 4-beta PSEC0001; lysosomal-associated transmembrane protein 4; Lysosomal-associated transmembrane protein 4B
Gene Aliases: C330023P13Rik; Laptm4b; LAPTM4beta
UniProt ID: (Mouse) Q91XQ6
Entrez Gene ID: (Mouse) 114128
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.