Invitrogen
Manufacturer Code:PA556206
Catalog # PIPA556206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LAP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteinylglycine-S-conjugate dipeptidase; Cytosol aminopeptidase; Cytosol aminopeptidase Leucine aminopeptidase Leucyl aminopeptidase Leucine aminopeptidase 3 LAP Proline aminopeptidase Prolyl aminopeptidase Peptidase S LAP3; epididymis secretory protein Li 106; LAP-3; Leucine aminopeptidase 3; Leucyl aminopeptidase; Peptidase S; Proline aminopeptidase; Prolyl aminopeptidase
Gene Aliases: HEL-S-106; LAP; LAP3; LAPEP; PEPS
UniProt ID: (Human) P28838
Entrez Gene ID: (Human) 51056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.