Proteintech
Manufacturer Code:146121AP150UL
Catalog # 50-172-9584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LAP3 Fusion Protein Ag6205 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteinylglycine-S-conjugate dipeptidase; Cytosol aminopeptidase; Cytosol aminopeptidase LAP LAP 3 LAP3 LAPEP leucine aminopeptidase 3 Leucyl aminopeptidase PEPS Peptidase S Proline aminopeptidase Prolyl aminopeptidase; epididymis secretory protein Li 106; LAP; LAP-3; Leucine aminopeptidase 3; Leucyl aminopeptidase; peptidase 7; Peptidase S; Proline aminopeptidase; Prolyl aminopeptidase
Gene Aliases: 2410015L10Rik; AA410100; HEL-S-106; LAP; LAP-3; LAP3; LAPEP; Pep-7; Pep-S; Pep7; PEPS
UniProt ID: (Human) P28838, (Rat) Q68FS4, (Mouse) Q9CPY7
Entrez Gene ID: (Human) 51056, (Rat) 289668, (Mouse) 66988
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.