Novus Biologicals
Manufacturer Code:NBP156294
Catalog # NBP156294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ANP32E(acidic (leucine-rich) nuclear phosphoprotein 32 family member E) The peptide sequence was selected from the middle region of ANP32E. Peptide sequence EDNEAPDSEEEDDEDGDEDDEEEEENEAGPPEGYEEEEEEEEEED |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acidic (leucine-rich) nuclear phosphoprotein 32 family member E LANP-like protein LANP-LLANPLacidic leucine-rich nuclear phosphoprotein 32 family member E leucine-rich acidic nuclear protein like MGC5350; acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; Acidic leucine-rich nuclear phosphoprotein 32 family member E; LANP-L; LANP-like protein; leucine-rich acidic nuclear protein like
Gene Aliases: ANP32E; LANP-L; LANPL
UniProt ID: (Human) Q9BTT0
Entrez Gene ID: (Human) 81611
Molecular Function:
enzyme modulator
phosphatase inhibitor
phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.