Invitrogen
Manufacturer Code:PA554301
Catalog # PIPA554301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LAMTOR4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Late endosomal/lysosomal adaptor and MAPK and MTOR activator 4; Ragulator complex protein LAMTOR4; Ragulator complex protein LAMTOR4, N-terminally processed; UPF0539 protein C7orf59; UPF0539 protein C7orf59 homolog
Gene Aliases: 0910001L09Rik; AV006840; C7orf59; LAMTOR4; RGD1309735
UniProt ID: (Human) Q0VGL1, (Mouse) Q8CF66
Entrez Gene ID: (Human) 389541, (Rat) 360776, (Mouse) 66096
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.