Invitrogen
Manufacturer Code:PA551792
Catalog # PIPA551792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LAMTOR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1; Lipid raft adaptor protein p18; p27kip1 releasing factor from RhoA; p27Kip1-releasing factor from RhoA; p27RF-Rho; PP7157 C11orf59 PDRO Ragulator1 p18 p27RF-Rho RhoA activator C11orf59 late endosomal/lysosomal adaptor and MAPK and MTOR activator 1 lipid raft adaptor protein p18 p27Kip1-releasing factor from RhoA p27kip1 releasing factor from RhoA protein associated with DRMs and endosomes ragulator complex protein LAMTOR1 ragulator complex protein PDRO; Protein associated with DRMs and endosomes; Ragulator complex protein LAMTOR1; ragulator complex protein PDRO; RhoA activator C11orf59
Gene Aliases: C11orf59; LAMTOR1; p18; p27RF-Rho; PDRO; PP7157; Ragulator1
UniProt ID: (Human) Q6IAA8
Entrez Gene ID: (Human) 55004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.