Invitrogen
Manufacturer Code:PA5114482
Catalog # PIPA5114482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rabbit polyclonal LAMTOR1 antibody was raised against a 17 amino acid peptide near the amino terminus of human LAMTOR1. The immunogen is located within the first 50 amino acids of LAMTOR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2400001E08Rik C11orf59 LAMTOR1 late endosomal/lysosomal adaptor and MAPK and MTOR activator 1 late endosomal/lysosomal adaptor MAPK and MTOR activator 1 lipid raft adaptor protein p18 p18 p27kip1 releasing factor from RhoA p27Kip1-releasing factor from RhoA p27RF Rho p27RF-Rho PDRO PP7157 Protein associated with DRMs and endosomes ragulator complex protein LAMTOR1 ragulator complex protein PDRO Ragulator1 RhoA activator C11orf59 RhoA activator C11orf59 homolog; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1; Lipid raft adaptor protein p18; p27kip1 releasing factor from RhoA; p27Kip1-releasing factor from RhoA; p27RF-Rho; Protein associated with DRMs and endosomes; Ragulator complex protein LAMTOR1; ragulator complex protein PDRO; RhoA activator C11orf59; RhoA activator C11orf59 homolog
Gene Aliases: 2400001E08Rik; C11orf59; LAMTOR1; p18; p27RF-Rho; PDRO; PP7157; Ragulator1
UniProt ID: (Human) Q6IAA8, (Rat) Q6P791, (Mouse) Q9CQ22
Entrez Gene ID: (Human) 55004, (Rat) 308869, (Mouse) 66508
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.