Proteintech
Manufacturer Code:145331AP150UL
Catalog # 50-172-9523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LAMR1RPSA Fusion Protein Ag6033 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 37 kDa laminin receptor; 37 kDa laminin receptor precursor; 37 kDa laminin receptor precursor {ECO:0000255|HAMAP-Rule:MF_03016}; 37 kDa oncofetal antigen; 37/67 kDa laminin receptor; 37/67 kDa laminin receptor 37LRP 40S ribosomal protein SA 67 kDa laminin receptor 67LR LAMBR Laminin receptor Laminin receptor 1 LamR LAMR1 LBP/p40 LRP LRP/LR NEM/1CHD4 p40 ribosomal protein SA RPSA; 37/67 kDa laminin receptor {ECO:0000255|HAMAP-Rule:MF_03016}; 37LRP; 37LRP {ECO:0000255|HAMAP-Rule:MF_03016}; 40S ribosomal protein SA; 40S ribosomal protein SA {ECO:0000255|HAMAP-Rule:MF_03016}; 67 kDa laminin receptor; 67 kDa laminin receptor {ECO:0000255|HAMAP-Rule:MF_03016}; 67LR; 67LR {ECO:0000255|HAMAP-Rule:MF_03016}; Colon carcinoma laminin-binding protein; Laminin receptor 1; laminin receptor 1 (67kD, ribosomal protein SA); laminin receptor 1 (ribosomal protein SA); laminin receptor 1 {ECO:0000255|HAMAP-Rule:MF_03016}; Laminin-binding protein precursor p40; laminin-binding protein precursor p40 {ECO:0000255|HAMAP-Rule:MF_03016}; LamR; lamR {ECO:0000255|HAMAP-Rule:MF_03016}; LBP/p40; LBP/p40 {ECO:0000255|HAMAP-Rule:MF_03016}; LRP/LR; LRP/LR {ECO:0000255|HAMAP-Rule:MF_03016}; Multidrug resistance-associated protein MGr1-Ag; NEM/1CHD4; OFA/iLRP; P40-3, functional; P40-8, functional; Small ribosomal subunit protein uS2
Gene Aliases: 37LRP; 67kDa; 67LR; AL022858; ICAS; LAMBR; Lamr; LAMR1; Lamrl1; LBP; LBP/p40; LRP; LRP/LR; MLR; NEM/1CHD4; P40; P40-3; P40-8; RPSA; SA
UniProt ID: (Human) P08865, (Rat) P38983, (Mouse) P14206
Entrez Gene ID: (Human) 3921, (Rat) 29236, (Mouse) 16785
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.