Invitrogen
Manufacturer Code:PA547576
Catalog # PIPA547576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human DC-LAMP Lumenal Domain Asp21-Thr381 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD208; CD208 DC LAMP DC-LAMP DCLAMP LAMP LAMP-3 TSC403 DC-lysosome-associated membrane glycoprotein lysosome-associated membrane glycoprotein 3; DC LAMP; DC-lysosome-associated membrane glycoprotein; LAMP-3; lysosomal-associated membrane protein 3; Lysosome-associated membrane glycoprotein 3; Protein TSC403
Gene Aliases: CD208; DC LAMP; DC-LAMP; DCLAMP; LAMP; LAMP-3; LAMP3; TSC403
UniProt ID: (Human) Q9UQV4
Entrez Gene ID: (Human) 27074
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.