Novus Biologicals
Manufacturer Code:NBP225155SS
Catalog # NBP225155SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 600 |
Immunogen | Full length human LAMP1 [UniProt# P11279] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD107 antigen-like family member A; CD107 antigen-like family member A CD107a CD107a antigen LAMP-1 LAMPA LGP120 lysosomal-associated membrane protein 1 lysosome-associated membrane glycoprotein 1 Lysosome-associated membrane protein 1; CD107a; LAMP-1; lysosomal-associated membrane protein 1; Lysosome-associated membrane glycoprotein 1; lysosome-associated membrane protein 1
Gene Aliases: CD107a; LAMP1; LAMPA; LGP120
UniProt ID: (Human) P11279
Entrez Gene ID: (Human) 3916
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.