Invitrogen
Manufacturer Code:PA576868
Catalog # PIPA576868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human LALBA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a-LACTAalfaLAAlpha-lactalbuminLactalbumin alphaLactose synthase B proteinLALBALALBA&lowbarHUMANLysozyme-like protein 7LYZL7; Alpha-lactalbumin; lactalbumin, alpha-; Lactose synthase B protein; lysozyme G; Lysozyme-like protein 7
Gene Aliases: LALBA; LYZG; LYZL7
UniProt ID: (Human) P00709, (Rat) P00714
Entrez Gene ID: (Human) 3906, (Rat) 24528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.