Invitrogen
Manufacturer Code:MA530627
Catalog # PIMA530627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 24 |
Immunogen | Recombit Human LAIR2 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD306; CD306 CD306 antigen CD306leukocyte-associated Ig-like receptor-2 LAIR-2 leukocyte-associated Ig-like receptor 2 leukocyte-associated immunoglobulin-like receptor 2 MGC71634; LAIR-2; leukocyte-associated Ig-like receptor 2; leukocyte-associated Ig-like receptor-2; Leukocyte-associated immunoglobulin-like receptor 2
Gene Aliases: CD306; LAIR2
UniProt ID: (Human) Q6ISS4
Entrez Gene ID: (Human) 3904
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.