Proteintech
Manufacturer Code:167831AP150UL
Catalog # 50-173-1176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LACTB2 Fusion Protein Ag10213 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta lactamase like protein 2 CGI 83 lactamase beta 2 LACTB2; Beta-lactamase-like protein 2; Endoribonuclease LACTB2; lactamase, beta 2; testicular secretory protein Li 23
Gene Aliases: CGI-83; E430032H21Rik; LACTB2
UniProt ID: (Human) Q53H82, (Rat) Q561R9, (Mouse) Q99KR3
Entrez Gene ID: (Human) 51110, (Rat) 297768, (Mouse) 212442
Molecular Function:
defense/immunity protein
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.