Invitrogen
Manufacturer Code:PA563342
Catalog # PIPA563342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human L3HYPDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L-3-hydroxyproline dehydratase (trans-); L-3-hydroxyproline dehydratase (trans-) L3HYPDH C14orf149; probable proline racemase; Trans-3-hydroxy-L-proline dehydratase; trans-3-hydroxyl-L-proline dehydratase; Trans-L-3-hydroxyproline dehydratase
Gene Aliases: C14orf149; L3HYPDH
UniProt ID: (Human) Q96EM0
Entrez Gene ID: (Human) 112849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.