Invitrogen
Manufacturer Code:MA511921
Catalog # PIMA511921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LPL4A.1 |
Immunogen | Human recombit L-plastin protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); epididymis secretory protein Li 37; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P) LCP-1 Lymphocyte cytosolic protein-1 (plasmin)bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)) plastin 2 plastin-2; LC64P; LCP-1; Lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; Plastin-2
Gene Aliases: CP64; HEL-S-37; L-PLASTIN; LC64P; LCP1; LPL; PLS2
UniProt ID: (Human) P13796
Entrez Gene ID: (Human) 3936
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.