Invitrogen
Manufacturer Code:PA582679
Catalog # PIPA582679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LCP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); CP64 L-PLASTIN LC64P PLS2; epididymis secretory protein Li 37; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); LC64P; LCP-1; Lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; Plastin-2
Gene Aliases: CP64; HEL-S-37; L-PLASTIN; LC64P; LCP1; LPL; PLS2
UniProt ID: (Human) P13796
Entrez Gene ID: (Human) 3936, (Rat) 306071
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.