Invitrogen
Manufacturer Code:PA544200
Catalog # PIPA544200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human LCP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa macrophage protein; bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); epididymis secretory protein Li 37; L-fimbrin; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (65 kDa macrophage protein) (PP65); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); LC64P; LCP-1; leucocyte-specific plastin 1; lymphocyte cytosolic plastin 1; Lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein 1 (L-plastin) CP64 DKFZp781A23186 FLJ25423 FLJ26114 FLJ39956 L-PLASTIN LC64P PLS2 LPL; Lymphocyte cytosolic protein-1 (plasmin); plastin 2; plastin 2, L; Plastin-2; pp65
Gene Aliases: AW536232; cb245; CP64; D14Ertd310e; fj24b12; HEL-S-37; L-PLASTIN; LC64P; LCP-1; LCP1; LPL; PLS2; pp65; wu:fj24b12
UniProt ID: (Human) P13796, (Mouse) Q61233
Entrez Gene ID: (Dog) 476921, (Horse) 100050663, (Rabbit) 100358782, (Bovine) 540990, (Human) 3936, (Guinea pig) 100717499, (Mouse) 18826, (Rat) 306071, (Zebrafish) 30583
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.