Novus Biologicals
Manufacturer Code:NBP245767
Catalog # NBP245767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7F12 |
Immunogen | Full length human recombinant protein of human KCNAB1(NP_751891) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AKR6A3 hKvb3 hKvBeta3 K(+) channel subunit beta-1 KCNA1Bpotassium channel beta 3 chain Kvb1.3 KV-BETA-1 potassium channel beta3 subunit potassium channel shaker chain beta 1a potassium voltage-gated channel beta subunit potassium voltage-gated channel shaker-related subfamily beta member 1 voltage-gated potassium channel beta-1 subunit voltage-gated potassium channel subunit beta-1; K(+) channel subunit beta-1; K+ channel Beta1a chain; Kv-beta-1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; Voltage-gated potassium channel subunit beta-1
Gene Aliases: AKR6A3; hKvb3; hKvBeta3; KCNA1B; KCNAB1; KV-BETA-1; Kvb1.3
UniProt ID: (Human) Q14722
Entrez Gene ID: (Human) 7881
Molecular Function: ion channel oxidoreductase potassium channel reductase transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.