Invitrogen
Manufacturer Code:MA516375
Catalog # PIMA516375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP130 |
Immunogen | Synthetic peptide corresponding to an internal region of human Ku80 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 86 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 80 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC85; CTCBF; DNA repair protein XRCC5; KARP-1 KARP1 KU80 KUB2 Ku86 NFIV 86 kDa subunit of Ku antigen ATP-dependent DNA helicase 2 subunit 2 ATP-dependent DNA helicase II 80 kDa subunit CTC box-binding factor 85 kDa subunit CTC85 CTCBF DNA repair protein XRCC5 Ku autoantigen 80kDa Ku86 autoantigen related protein 1 TLAA X-ray; Ku autoantigen, 80kDa; Ku80; Ku86; Ku86 autoantigen related protein 1; Lupus Ku autoantigen protein p86; Nuclear factor IV; Thyroid-lupus autoantigen; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair cross-complementing protein 5
Gene Aliases: G22P2; KARP-1; KARP1; KU80; Ku86; KUB2; NFIV; XRCC5
UniProt ID: (Human) P13010
Entrez Gene ID: (Human) 7520
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.