Invitrogen
Manufacturer Code:PA517454
Catalog # PIPA517454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the carboxy terminus of human Ku80 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 86 kDa subunit of Ku antigen; 86 kDa subunit of Ku antigen ATP-dependent DNA helicase 2 subunit 2 ATP-dependent DNA helicase II 80 kDa subunit CTC box-binding factor 85 kDa subunit CTC85 CTCBF DNA repair protein XRCC5 Ku autoantigen 80kDa Ku86 autoantigen related protein 1 OTTHUMP00000164061 OTTHUMP00000206791 TLAA X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining Ku autoantigen 80kD) X-ray repair cross-complementing protein 5 X-ray repair complementing de; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 80 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC85; CTCBF; DNA repair protein XRCC5; Ku autoantigen protein p86 homolog; Ku autoantigen, 80kD); Ku autoantigen, 80kDa; Ku p80; Ku80; Ku86; Ku86 autoantigen related protein 1; Lupus Ku autoantigen protein p86; Nuclear factor IV; Thyroid-lupus autoantigen; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair cross complementation (double-strand-break rejoining; X-ray repair cross complementation (double-strand-break rejoining; Ku autoantigen, 80kD); X-ray repair cross-complementing protein 5
Gene Aliases: AI314015; G22P2; KARP-1; KARP1; KU80; Ku86; KUB2; Kup80; NFIV; XRCC5
UniProt ID: (Human) P13010, (Mouse) P27641
Entrez Gene ID: (Human) 7520, (Rat) 363247, (Mouse) 22596
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.