Invitrogen
Manufacturer Code:MA512938
Catalog # PIMA512938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Gel Shift (GS) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 162 |
Immunogen | Human B cell nuclei from plasmacytoid 2p68 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Ku70 + Ku80 Monoclonal antibody MA5-12938 from Thermo Scientific™ specifically detects Ku70 + Ku80 in Human, Mouse, Non-human primate and Xenopus laevis samples. It is validated for Flow Cytometry, Gel Shift, Immunocytochemistry, Immunofluorescence and Immunohistochemistry (Paraffin).
For Research Use Only
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; 86 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC75; CTC85; CTCBF; DNA repair protein XRCC5; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen protein p70 homolog; Ku autoantigen protein p86 homolog; Ku autoantigen, 70kDa; Ku autoantigen, 80kDa; Ku p70; Ku p80; Ku70; Ku80; Ku86; Ku86 autoantigen related protein 1; Lupus Ku autoantigen protein p70; Lupus Ku autoantigen protein p86; Nuclear factor IV; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross-complementing protein 5; X-ray repair cross-complementing protein 6; XRCC6 KU70 Xrcc6 G22P1 X-ray repair cross-complementing protein 6 5&apos-deoxyribose-5-phosphate lyase Ku70 5&apos-dRP/AP lyase Ku70 ATP-dependent DNA helicase 2 subunit 1 ATP-dependent DNA helicase II 70 kDa subunit CTC box-binding factor 75 k
Gene Aliases: 70kDa; AI314015; CTC75; CTCBF; G22P1; G22P2; KARP-1; KARP1; KU70; KU80; Ku86; KUB2; ML8; NFIV; TLAA; XELAEV_18023498mg; XRCC5; XRCC6
UniProt ID: (Human) P13010, (Human) P12956, (Mouse) P27641, (Mouse) P23475
Entrez Gene ID: (Human) 7520, (Human) 2547, (Mouse) 22596, (Mouse) 14375, (Xenopus) 398357
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.