Novus Biologicals
Manufacturer Code:NBP266920
Catalog # NBP266920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM61-31 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-deoxyribose-5-phosphate lyase Ku70 5'-dRP lyase Ku70 70 kDa subunit of Ku antigen ATP-dependent DNA helicase 2 subunit 1 ATP-dependent DNA helicase II 70 kDa subunit CTC box binding factor 75 kDa subunit CTC box-binding factor 75 kDa subunit CTC75 CTCBF D22S731 EC 3.6.4.- EC 4.2.99.- G22P1Ku70 Ku autoantigen p70 subunit Ku autoantigen 70kDa KU70 ML870 kDa subunit Thyroid-lupus autoantigen thyroid-lupus autoantigen p70 TLAAD22S671 X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6 X-ray repair cross-complementing protein 6; 5'-dRP lyase Ku70; 70 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen, 70kDa; Ku70; Lupus Ku autoantigen protein p70; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross-complementing protein 6
Gene Aliases: CTC75; CTCBF; G22P1; KU70; ML8; TLAA; XRCC6
UniProt ID: (Human) P12956
Entrez Gene ID: (Human) 2547
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.