Invitrogen
Manufacturer Code:PA599488
Catalog # PIPA599488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Ku70/80 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-deoxyribose-5-phosphate lyase Ku70 5'-dRP lyase Ku70 5'-dRP/AP lyase Ku70 70 kDa subunit of Ku antigen 70kDa 86 kDa subunit of Ku antigen AI314015 ATP-dependent DNA helicase 2 subunit 1 ATP-dependent DNA helicase 2 subunit 2 ATP-dependent DNA helicase II 70 kDa subunit ATP-dependent DNA helicase II 80 kDa subunit ATP-dependent DNA helicase II 70 kDa subunit CTC box binding factor 75 kDa subunit CTC box-binding factor 75 kDa subunit CTC box-binding factor 85 kDa subunit CTC75 CTC85 CTCBF DNA repair protein XRCC5 DNA repair protein XRCC6 G22P1 G22P2 KARP1 KARP-1 Ku autoantigen p70 subunit ku autoantigen protein p70 homolog Ku autoantigen protein p86 homolog Ku autoantigen 70kDa Ku autoantigen 80kD) Ku autoantigen 80kDa Ku p70 Ku p80 Ku70 Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa) Ku80 Ku86 Ku86 autoantigen related protein 1 KUB2 Kup70 Kup80 Lupus Ku autoantigen protein p70 lupus Ku autoantigen protein p86 ML8 NFIV Nuclear factor IV thyroid autoantigen thyroid autoantigen 70 kDa thyroid autoantigen 70kD (Ku antigen) thyroid autoantigen 70kDa (Ku antigen) Thyroid-lupus autoantigen thyroid-lupus autoantigen p70 TLAA XELAEV_18023498mg XELAEV_18044412mg X-ray repair complementing defective repair in Chinese hamster cells 5 X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining) X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining Ku autoantigen 80kDa) X-ray repair complementing defective repair in Chinese hamster cells 5 L homeolog X-ray repair complementing defective repair in Chinese hamster cells 6 X-ray repair complementing defective repair in Chinese hamster cells 6 L homeolog X-ray repair cross complementation (double-strand-break rejoining X-ray repair cross complementation (double-strand-break rejoining Ku autoantigen 80kD) X-ray repair cross complementing 5 X-ray repair cross complementing 6 X-ray repair cross-complementing protein 5 X-ray repair cross-complementing protein 6 xrcc5 xrcc5.L xrcc5-a XRCC6 xrcc6.L; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; 86 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC75; CTC85; CTCBF; DNA repair protein XRCC5; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen protein p70 homolog; Ku autoantigen protein p86 homolog; Ku autoantigen, 70kDa; Ku autoantigen, 80kD); Ku autoantigen, 80kDa; Ku p70; Ku p80; Ku70; Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa); Ku80; Ku86; Ku86 autoantigen related protein 1; Lupus Ku autoantigen protein p70; Lupus Ku autoantigen protein p86; Nuclear factor IV; thyroid autoantigen; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross complementation (double-strand-break rejoining; X-ray repair cross complementation (double-strand-break rejoining; Ku autoantigen, 80kD); X-ray repair cross-complementing protein 5; X-ray repair cross-complementing protein 6
Gene Aliases: 70kDa; AI314015; CTC75; CTCBF; G22P1; G22P2; KARP-1; KARP1; KU70; KU80; Ku86; KUB2; Kup70; Kup80; ML8; NFIV; TLAA; XRCC5; XRCC6
UniProt ID: (Human) P13010, (Human) P12956, (Mouse) P27641, (Mouse) P23475
Entrez Gene ID: (Human) 7520, (Human) 2547, (Rat) 363247, (Mouse) 22596, (Rat) 25019, (Mouse) 14375
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.