Invitrogen
Manufacturer Code:PA525915
Catalog # PIPA525915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 432-461 amino acids from the central region of human Ku70 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; CTC75 CTCBF G22P1 KU70 ML8 TLAA 5&prime-dRP lyase Ku70 5&prime-deoxyribose-5-phosphate lyase Ku70 70 kDa subunit of Ku antigen ATP-dependent DNA helicase 2 subunit 1 ATP-dependent DNA helicase II 70 kDa subunit ATP-dependent DNA helicase II 70 kDa subunit CTC box binding factor 75 kDa subunit CT; CTCBF; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen protein p70 homolog; Ku autoantigen, 70kDa; Ku p70; Ku70; Lupus Ku autoantigen protein p70; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross-complementing protein 6
Gene Aliases: 70kDa; CTC75; CTCBF; G22P1; KU70; ML8; TLAA; XRCC6
UniProt ID: (Human) P12956, (Mouse) P23475
Entrez Gene ID: (Human) 2547, (Mouse) 14375
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.