R&D Systems
Manufacturer Code:FAB7099U100UG
Catalog # FAB7099U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 854929 |
Immunogen | Chinese hamster ovary cell line CHO derived recombinant mouse Klre-1 Lys94-Lys226 Accession # NP_705818 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: CD94; CD94 antigen; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; KP43; Natural killer cells antigen CD94; NK cell receptor
Gene Aliases: CD94; KLRD1
UniProt ID: (Human) O43321
Entrez Gene ID: (Human) 3824
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.