R&D Systems
Manufacturer Code:MAB5889
Catalog # MAB5889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 885935 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Klotho beta Phe53-Leu997 Accession # Q86Z14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-klotho; beta-klotho betaKlotho BKL KLB klotho beta klotho beta like klotho beta-like protein MGC142213; betaKlotho; BKL; klotho beta like; Klotho beta-like protein
Gene Aliases: BKL; KLB
UniProt ID: (Human) Q86Z14
Entrez Gene ID: (Human) 152831
Molecular Function:
glucosidase
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.