Invitrogen
Manufacturer Code:PA542022
Catalog # PIPA542022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2°-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Klotho; Klotho peptide; secreted form of Klotho protein
Gene Aliases: alpha-kl; KL
UniProt ID: (Human) Q9UEF7, (Rat) Q9Z2Y9, (Mouse) O35082
Entrez Gene ID: (Horse) 100064836, (Bovine) 784635, (Dog) 100685604, (Rabbit) 100356074, (Human) 9365, (Guinea pig) 100714519, (Rat) 83504, (Mouse) 16591
Molecular Function: glucosidase glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.