Abnova
Manufacturer Code:MAB5967
Catalog # 89-112-592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 2B8 |
Immunogen | Native purified Kit from murine bone marrow mast cells. |
Conjugate | APC-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against native Kit.
For Research Use Only
Protein Aliases: belly-spot; Bs; c-KIT; c-kit proto-oncogene protein; CD117; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; Mast/stem cell growth factor receptor Kit; Proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; SCFR; SCO1; SCO5; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; Tyrosine-protein kinase Kit; W
Gene Aliases: Bs; c-KIT; CD117; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; Kit; SCO1; SCO5; Sl; SOW3; Ssm; Tr-kit; W
UniProt ID: (Mouse) P05532
Entrez Gene ID: (Mouse) 16590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.