Novus Biologicals
Manufacturer Code:NBP238788
Catalog # NBP238788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1867MGC126824 kin of IRRE like 3 (Drosophila) Kin of irregular chiasm-like protein 3 kin of IRRE-like protein 3 KIRREPRO4502 MRD4 NEPH2MGC126850 nephrin-like 2 Nephrin-like protein 2; Kin of IRRE-like protein 3; Kin of irregular chiasm-like protein 3; nephrin-like 2; Nephrin-like protein 2; Processed kin of IRRE-like protein 3
Gene Aliases: KIAA1867; KIRRE; KIRREL3; MRD4; NEPH2; PRO4502; UNQ5923/PRO4502/PRO19814
UniProt ID: (Human) Q8IZU9
Entrez Gene ID: (Human) 84623
Molecular Function: cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.