Invitrogen
Manufacturer Code:PA568682
Catalog # PIPA568682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat KCNJ16 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BIR9; Inward rectifier K(+) channel Kir5.1; inward rectifier K+ channel KIR5.1; Inward rectifier potassium channel 16; inwardly-rectifying potassium channel Kir5.1; Potassium channel, inwardly rectifying subfamily J member 16; potassium channel, inwardly rectifying subfamily J, member 16; potassium inwardly-rectifying channel J16; Potassium inwardly-rectifying channel subfamily J member 16 BIR9 KIR5.1 MGC33717; potassium inwardly-rectifying channel, subfamily J, member 16; potassium voltage-gated channel subfamily J member 16
Gene Aliases: 6430410F18Rik; AI132396; BIR9; KCNJ16; KIR5.1
UniProt ID: (Human) Q9NPI9, (Mouse) Q9Z307
Entrez Gene ID: (Rabbit) 100355482, (Dog) 611045, (Horse) 100053003, (Bovine) 613890, (Human) 3773, (Mouse) 16517, (Rat) 29719, (Guinea pig) 100379597
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.