Proteintech
Manufacturer Code:126961AP150UL
Catalog # 50-172-8181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Kir5.1 Fusion Protein Ag3405 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BIR9; BIR9 KCNJ16 KIR5.1; Inward rectifier K(+) channel Kir5.1; inward rectifier K+ channel KIR5.1; Inward rectifier potassium channel 16; Potassium channel, inwardly rectifying subfamily J member 16; potassium channel, inwardly rectifying subfamily J, member 16; potassium inwardly-rectifying channel, subfamily J, member 16
Gene Aliases: 6430410F18Rik; AI132396; BIR9; KCNJ16; KIR5.1
UniProt ID: (Human) Q9NPI9, (Mouse) Q9Z307
Entrez Gene ID: (Human) 3773, (Mouse) 16517, (Rat) 29719
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.