Invitrogen
Manufacturer Code:PA568685
Catalog # PIPA568685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human KCNJ12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-sensitive inward rectifier potassium channel 12; ATP-sensitive inward rectifier potassium channel 12 Inward rectifier K(+) channel Kir22 IRK-2 Inward rectifier K(+) channel Kir22v Potassium channel inwardly rectifying subfamily J member 12 KCNJ12 IRK2 KCNJN1; atrium potassium channel IRK; DirK; Inward rectifier K(+) channel Kir2.2; Inward rectifier K(+) channel Kir2.2v; inward rectifier K(+) channel Kir2.6; inwardly-rectifying potassium channel Kir2.2; IRK-2; Potassium channel, inwardly rectifying subfamily J member 12; potassium channel, inwardly rectifying subfamily J, member 12; potassium inwardly-rectifying channel J12; potassium inwardly-rectifying channel, subfamily J, inhibitor 1; potassium inwardly-rectifying channel, subfamily J, member 12; potassium voltage-gated channel subfamily J member 12; subfamily J, member 12
Gene Aliases: hIRK; hIRK1; hkir2.2x; IRK; IRK-2; IRK2; KCNJ12; kcnj12x; KCNJN1; Kir2.1; KIR2.2; Kir2.2v; MB-IRK2
UniProt ID: (Human) Q14500, (Bovine) Q4TZY1, (Mouse) P52187
Entrez Gene ID: (Horse) 100147042, (Human) 3768, (Dog) 403760, (Bovine) 538479, (Guinea pig) 100169698, (Mouse) 16515, (Rat) 117052
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.