Novus Biologicals
Manufacturer Code:NBP287693
Catalog # NB035315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human Kir2.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-sensitive inward rectifier potassium channel 12; ATP-sensitive inward rectifier potassium channel 12 hIRK hIRK1 hkir2.2x Inward rectifier K(+) channel Kir2.2 Inward rectifier K(+) channel Kir2.2v inward rectifier K(+) channel Kir2.6 IRK2IRK-2 kcnj12x KCNJN1FLJ14167 Kir2.2 Kir2.2v Potassium channel inwardly rectifying subfamily J member 12 potassium inwardly-rectifying channel subfamily J inhibitor 1 potassium inwardly-rectifying channel subfamily J member 12; Inward rectifier K(+) channel Kir2.2; Inward rectifier K(+) channel Kir2.2v; inward rectifier K(+) channel Kir2.6; IRK-2; Potassium channel, inwardly rectifying subfamily J member 12; potassium channel, inwardly rectifying subfamily J, member 12; potassium inwardly-rectifying channel, subfamily J, inhibitor 1
Gene Aliases: hIRK; hIRK1; hkir2.2x; IRK-2; IRK2; KCNJ12; kcnj12x; KCNJN1; Kir2.2; Kir2.2v
UniProt ID: (Human) Q14500
Entrez Gene ID: (Human) 3768
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.