Proteintech
Manufacturer Code:661231IG150UL
Catalog # 50-173-6559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10E8D11 |
Immunogen | Kininogen 1 Fusion Protein Ag19233 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2-thiol proteinase inhibitor; BDK Fitzgerald factor HMWK Ile Ser Bradykinin Bradykinin Kallidin I Lysyl bradykinin kininogen 1 KNG KNG1; Bradykinin; Fitzgerald factor; High molecular weight kininogen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; Kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Low molecular weight growth-promoting factor; Lysyl-bradykinin; T-kinin; Williams-Fitzgerald-Flaujeac factor
Gene Aliases: BDK; BK; KNG; KNG1
UniProt ID: (Human) P01042
Entrez Gene ID: (Human) 3827
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.