Novus Biologicals
Manufacturer Code:NBP158177
Catalog # NBP158177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to KIF5B (kinesin family member 5B) The peptide sequence was selected from the N terminal of KIF5B. Peptide sequence CNIKVMCRFRPLNESEVNRGDKYIAKFQGEDTVVIASKPYAFDRVFQSST. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Conventional kinesin heavy chain; Conventional kinesin heavy chain kinesin family member 5B kinesin-1 heavy chain KNS1kinesin 1 (110-120kD) KNSKINH Ubiquitous kinesin heavy chain UKHCkinesin heavy chain; epididymis secretory protein Li 61; kinesin 1 (110-120kD); kinesin heavy chain; Kinesin-1 heavy chain; Ubiquitous kinesin heavy chain; UKHC
Gene Aliases: HEL-S-61; KIF5B; KINH; KNS; KNS1; UKHC
UniProt ID: (Human) P33176
Entrez Gene ID: (Human) 3799
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.