Invitrogen
Manufacturer Code:MA190667
Catalog # PIMA190667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | P220 1F8/3 |
Immunogen | Recombit Kidins220/ARMS (last 347 amino acids at the carboxy terminus) protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat-rich membrane spanning protein; Ankyrin repeat-rich membrane-spanning protein; ARMS; kinase D-interacting substance 220; kinase D-interacting substance of 220 kDa; Kinase D-interacting substrate of 220 kDa
Gene Aliases: ARMS; Kidins220
UniProt ID: (Rat) Q9EQG6
Entrez Gene ID: (Rat) 116478
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.