Invitrogen
Manufacturer Code:PA553339
Catalog # PIPA553339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Kiaa0319 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dyslexia susceptibility 2; Dyslexia-associated protein KIAA0319; mKIAA0319 similar to KIAA0319 gene product 4930451E12Rik KIAA0319; neuronal migration
Gene Aliases: DYLX2; DYX2; KIAA0319; NMIG
UniProt ID: (Human) Q5VV43
Entrez Gene ID: (Human) 9856
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.