Novus Biologicals
Manufacturer Code:NB500170
Catalog # NB500170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67 between amino acids: 1218-1232 1582-1596 2310-2324 [Uniprot: P46013]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen identified by monoclonal antibody Ki-67; antigen identified by monoclonal Ki-67 antigen KI-67 Antigen Ki67 antiKi67 Ki67 Ki-67 Ki67 ihc Ki-67 ihc Ki67 mouse Ki-67 mouse Ki67 western blot Ki-67 western blot KIA Marker Of Proliferation Ki-67 MIB- MIB-1 PPP1R105 Proliferation Marker Protein Ki-67 proliferation-related Ki-67 antigen Protein Phosphatase 1 Regulatory Subunit 105; Antigen KI-67; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105
Gene Aliases: KIA; MIB-; MIB-1; MKI67; PPP1R105
UniProt ID: (Human) P46013
Entrez Gene ID: (Human) 4288
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.