Novus Biologicals
Manufacturer Code:NBP271069
Catalog # NBP271069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D8 |
Immunogen | Full length human recombinant protein of human KHK (NP_000212) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.1.3 Hepatic fructokinase ketohexokinase ketohexokinase (fructokinase); fructokinase; Hepatic fructokinase; Ketohexokinase; ketohexokinase (fructokinase); testicular tissue protein Li 102
Gene Aliases: KHK
UniProt ID: (Human) P50053
Entrez Gene ID: (Human) 3795
Molecular Function:
carbohydrate kinase
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.