Novus Biologicals
Manufacturer Code:NBP271069PEATT594
Catalog # N271069P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D8 |
Immunogen | Full length recombinant protein of human Ketohexokinase (NP_000212) produced in HEK293T cells. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.1.3 Hepatic fructokinase ketohexokinase ketohexokinase (fructokinase); fructokinase; Hepatic fructokinase; Ketohexokinase; ketohexokinase (fructokinase); testicular tissue protein Li 102
Gene Aliases: KHK
UniProt ID: (Human) P50053
Entrez Gene ID: (Human) 3795
Molecular Function: carbohydrate kinase kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.