Novus Biologicals
Manufacturer Code:NBP211755AF647
Catalog # NB211755647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0435-12X3 |
Immunogen | Human Blood Group Kell Antigen |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CD238; CD238 CD238 antigen EC 3.4.24.- ECE3 Kell blood group kell blood group antigen kell blood group glycoprotein Kell blood group metalloendopeptidase Kell blood group metallo-endopeptidase; kell blood group antigen; Kell blood group glycoprotein
Gene Aliases: CD238; ECE3; KEL
UniProt ID: (Human) P23276
Entrez Gene ID: (Human) 3792
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.