Novus Biologicals
Manufacturer Code:NBP237431
Catalog # NBP237431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G4B10 |
Immunogen | Purified recombinant fragment of human KEAP1 (AA: 380-624 ) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytosolic inhibitor of Nrf2; INrf2; INRF2 INrf2MGC9454 kelch-like ECH-associated protein 1 Kelch-like protein 19 KIAA0132MGC4407 KLHL19Cytosolic inhibitor of Nrf2 MGC10630 MGC1114 MGC20887; Kelch-like ECH-associated protein 1; kelch-like family member 19; Kelch-like protein 19
Gene Aliases: INRF2; KEAP1; KIAA0132; KLHL19
UniProt ID: (Human) Q14145
Entrez Gene ID: (Human) 9817
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.