Abnova
Manufacturer Code:MAB11736
Catalog # 89-109-870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S18A-3 |
Immunogen | Recombinant fusion protein corresponding to amino acids 45-167 at extracellular domain of mouse Kcnmb4. |
Conjugate | ATTO 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial synthetic protein of Kcnmb4 .
For Research Use Only
Protein Aliases: 1700058G18Rik; 2900045G12Rik; BK channel subunit beta-4; BKbeta4; Calcium-activated potassium channel subunit beta-4; Calcium-activated potassium channel, subfamily M subunit beta-4; Charybdotoxin receptor subunit beta-4; K(VCA)beta-4; large conductance calcium-dependent potassium ion channel beta 4 subunit; Maxi K channel subunit beta-4; MGC143660; Slo-beta-4; Slowpoke beta 4
Gene Aliases: 1700058G18Rik; 2900045G12Rik; Kcnmb4
UniProt ID: (Mouse) Q9JIN6
Entrez Gene ID: (Mouse) 58802
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.