Abnova
Manufacturer Code:MAB19746
Catalog # 89-339-031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S22 |
Immunogen | Recombinant protein corresponding to amino acids 690-1196 at C-terminus of mouse Kcnma1 |
Conjugate | ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Mouse Monoclonal Antibody.
Protein Aliases: BK channel; BK channel alpha subunit; BKCA alpha; Calcium-activated potassium channel subunit alpha-1; Calcium-activated potassium channel, subfamily M subunit alpha-1; K(VCA)alpha; KCa1.1; Maxi K channel; MaxiK; MaxiK Slo Slo1 mSlo mSlo1; mSlo1; Slo homolog; Slo-alpha; Slo1; Slowpoke homolog
Gene Aliases: 5730414M22Rik; BKCa; Kcnma; Kcnma1; MaxiK; mSlo; mSlo1; Slo; Slo1
UniProt ID: (Mouse) Q08460
Entrez Gene ID: (Mouse) 16531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.