Abnova
Manufacturer Code:MAB18698
Catalog # 89-338-623
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S374-48 |
Immunogen | Recombinant protein corresponding to amino acids 251-411 at C-terminus of rat Kcnk3. |
Conjugate | ATTO700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensitive potassium channel protein TASK-1; Potassium channel subfamily K member 3; potassium channel, subfamily K, member 3; rTASK; Task-1; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; Two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Gene Aliases: Kcnk3; rTASK; Task; Task-1; Task1
UniProt ID: (Rat) O54912
Entrez Gene ID: (Rat) 29553
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.