Abnova
Manufacturer Code:MAB8581
Catalog # 89-113-511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S55-82 |
Immunogen | Recombinant protein corresponding to full length of rat Kcnip1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant Kcnip1.
For Research Use Only
Protein Aliases: A-type potassium channel modulatory protein 1; KChIP1; Kv channel-interacting protein 1; Potassium channel-interacting protein 1
Gene Aliases: Kchip1; Kcnip1
UniProt ID: (Rat) Q8R426
Entrez Gene ID: (Rat) 65023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.