Abnova
Manufacturer Code:MAB16514
Catalog # 89-999-455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | G9 |
Immunogen | Recombinant protein corresponding to full length rat Kcnip1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A-type potassium channel modulatory protein 1; KChIP1; Kv channel-interacting protein 1; Potassium channel-interacting protein 1
Gene Aliases: Kchip1; Kcnip1
UniProt ID: (Rat) Q8R426
Entrez Gene ID: (Rat) 65023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.