Abnova
Manufacturer Code:MAB11814
Catalog # 89-109-948
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S4-11 |
Immunogen | Recombinant GST fusion protein corresponding to amino acids of 509-853 of rat Kcnb1. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant Kcnb1.
For Research Use Only
Protein Aliases: Delayed rectifier potassium channel 1; DRK1; DRK1PC; Kcr1-1; Kv2.1; potassium channel Kv2.1; potassium voltage gated channel, Shab-related subfamily, member 1; Potassium voltage-gated channel subfamily B member 1; Shab; Voltage-gated potassium channel subunit Kv2.1
Gene Aliases: DRK1PC; Kcnb1; Kcr1-1; Kv2.1; Shab
UniProt ID: (Rat) P15387
Entrez Gene ID: (Rat) 25736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.