Abnova
Manufacturer Code:MAB19402
Catalog # 89-338-906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | G6 |
Immunogen | Recombinant protein corresponding to full length rat Kcnab2 |
Conjugate | ATTO 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Mouse Monoclonal Antibody.
Protein Aliases: F5 I2rf5 Kcnb3; K(+) channel subunit beta-2; Kv-beta-2; Neuroimmune protein F5; potassium voltage gated channel, shaker related subfamily, beta member 2; Voltage-gated potassium channel subunit beta-2
Gene Aliases: Ckbeta2; F5; I2rf5; Kcnab2; Kcnb3; kv-beta-2
UniProt ID: (Mouse) P62482
Entrez Gene ID: (Mouse) 16498
Molecular Function: ion channel oxidoreductase potassium channel reductase transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.