Abnova
Manufacturer Code:MAB16501
Catalog # 89-999-442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | XF1 |
Immunogen | A synthetic peptide corresponding to amino acids 7-28 at N-terminus of rat Kcnab1. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: K(+) channel subunit beta-1; Kv-beta-1; KvBeta3; MGC105463; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; Voltage-gated potassium channel subunit beta-1
Gene Aliases: Kcnab1; Kvb1; KvBeta3
UniProt ID: (Rat) P63144
Entrez Gene ID: (Rat) 29737
Molecular Function:
ion channel
oxidoreductase
potassium channel
reductase
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.